missing translation for 'onlineSavingsMsg'
Learn More

(S)-Amlodipine, TRC

Product Code. 30006803
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006803 100 mg 100mg
30137095 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30006803 Supplier Toronto Research Chemicals Supplier No. TRCA633491100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103129-82-4
Molecular Formula C20H25ClN2O5
Molecular Weight (g/mol) 408.88
Synonym 3,5-Pyridinedicarboxylic acid, 2-[(2-aminoethoxy)methyl]-4-(2-chlorophenyl)-1,4-dihydro-6-methyl-, 3-ethyl 5-methyl ester, (4S)-, 3,5-Pyridinedicarboxylic acid, 2-[(2-aminoethoxy)methyl]-4-(2-chlorophenyl)-1,4-dihydro-6-methyl-, 3-ethyl 5-methyl ester, (S)-, (-)-Amlodipine, (S)-(-)-Amlodipine, (S)-Amlodipine, Levamlodipine, Levoamlodipine, Lodien, l-Amlodipine
IUPAC Name 3-O-ethyl 5-O-methyl (4S)-2-(2-aminoethoxymethyl)-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CCOC(=O)C1=C(COCCN)NC(=C([C@@H]1c2ccccc2Cl)C(=O)OC)C

Specifications

Analyte or Component Names (S)-Amlodipine
Shipping Condition Room Temperature
InChI Formula InChI=1S/C20H25ClN2O5/c1-4-28-20(25)18-15(11-27-10-9-22)23-12(2)16(19(24)26-3)17(18)13-7-5-6-8-14(13)21/h5-8,17,23H,4,9-11,22H2,1-3H3/t17-/m0/s1
Solution Type Neat
Industry Type Forensics
Quantity 100 mg
Formula Weight 408.1452
Percent Purity >95%
Chemical Name or Material (S)-Amlodipine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.