missing translation for 'onlineSavingsMsg'
Learn More

Psoralen, MedChemExpress

Product Code. 16448966
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16448966

Brand: Medchem Express HYN0053

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Psoralen (Ficusin) is a coumarin isolated from the seeds of Fructus Psoraleae. Psoralen exhibits a wide range of biological properties, including anti-cancer, antioxidant, antidepressant, anticancer, antibacterial, and antiviral, et al.

Psoralen (Ficusin) is a coumarin isolated from the seeds of Fructus Psoraleae. Psoralen exhibits a wide range of biological properties, including anti-cancer, antioxidant, antidepressant, anticancer, antibacterial, and antiviral, et al.

Specifications

Chemical Name or Material Psoralen
Physical Form Solid
CAS 66-97-7
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C11H6O3
For Use With (Application) Cancer-programmed cell death
Synonym Ficusin
Solubility Information DMSO : 100 mg/mL (537.17 mM; Need ultrasonic) ∣H2O : 1 mg/mL (5.37 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C1C=CC2=CC3=C(OC=C3)C=C2O1
Molecular Weight (g/mol) 186.16
Formula Weight 186.16
Percent Purity 99.67%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.