missing translation for 'onlineSavingsMsg'
Learn More

Poliumoside, MedChemExpress

Product Code. 16408966
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16408966

Brand: Medchem Express HYN0033

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Poliumoside, a caffeoylated phenylpropanoid glycoside, is isolated from Brandisia hancei stems and leaves. Poliumoside is an advanced glycation end product (AGE) formation and rat lens aldose reductase (RLAR) inhibitor, with IC50s of 19.69 and 8.47 μM, respectively. Poliumoside also has antiinflammatory and antioxidant activity.

Poliumoside, a caffeoylated phenylpropanoid glycoside, is isolated from Brandisia hancei stems and leaves. Poliumoside is an advanced glycation end product (AGE) formation and rat lens aldose reductase (RLAR) inhibitor, with IC50s of 19.69 and 8.47 μM, respectively. Poliumoside also has antiinflammatory and antioxidant activity.

Specifications

Chemical Name or Material Poliumoside
CAS 94079-81-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C35H46O19
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (129.75 mM; Need ultrasonic) ∣Ethanol : 50 mg/mL (64.87 mM; Need ultrasonic)
SMILES OC1=C(O)C=CC(CCOC2C(C(OC3OC(C)C(O)C(O)C3O)C(OC(/C=C/C4=CC=C(O)C(O)=C4)=O)C(COC5OC(C)C(O)C(O)C5O)O2)O)=C1
Molecular Weight (g/mol) 770.73
Formula Weight 770.73
Percent Purity 99.8%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.