missing translation for 'onlineSavingsMsg'
Learn More

Pheniramine N-Oxide, Mikromol™

Product Code. 30050603
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30050603

Brand: Mikromol MM0059.04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Pheniramine N-Oxide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pheniramine N-Oxide
CAS 12656-98-3
Analyte or Component Names Pheniramine N-Oxide
Shipping Condition Room Temperature
Molecular Formula C16 H20 N2 O
Certifications/Compliance ISO 9001
Synonym Pheniramine N-oxide, 1-Phenyl-1-(2-pyridyl)-3-dimethylaminopropane N-Oxide, N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine N-oxide
InChI Formula InChI=1S/C16H20N2O/c1-18(2,19)13-11-15(14-8-4-3-5-9-14)16-10-6-7-12-17-16/h3-10,12,15H,11,13H2,1-2H3
SMILES C[N+](C)([O-])CCC(c1ccccc1)c2ccccn2
IUPAC Name N,N-dimethyl-3-phenyl-3-pyridin-2-ylpropan-1-amine oxide
Molecular Weight (g/mol) 256.34
Quantity 100 mg
Formula Weight 256.1576 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.