missing translation for 'onlineSavingsMsg'
Learn More

Naltrexone, TRC

Product Code. 17920405
Change view
Click to view available options
Quantity:
100 mg
1 g
2.5 g
Unit Size:
100mg
1g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17920405 100 mg 100mg
30134935 2.5 g 2.5g
30133799 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17920405 Supplier Toronto Research Chemicals Supplier No. TRCN285745100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16590-41-3
Molecular Formula C20 H23 N O4
Molecular Weight (g/mol) 341.4
Synonym (5α)-17-(Cyclopropylmethyl)-4,5-epoxy-3,14-dihydroxymorphinan-6-one, Morphinan-6-one, 17-(cyclopropylmethyl)-4,5α-epoxy-3,14-dihydroxy- (8CI), (-)-Naltrexone, 1-N-Cyclopropylmethyl-7,8-dihydro-14-hydroxynormorphinone, Depotrex, EN 1639, N-Cyclopropylmethylnoroxymorphone, Naltrel, Naltrexone, Nemexin, Trexonil, UM 792, Vivitrex, Vivitrol
IUPAC Name (4R,4aS,7aR,12bS)-3-(cyclopropylmethyl)-4a,9-dihydroxy-2,4,5,6,7a,13-hexahydro-1H-4,12-methanobenzofuro[3,2-e]isoquinoline-7-one
SMILES Oc1ccc2C[C@H]3N(CC4CC4)CC[C@@]56[C@@H](Oc1c25)C(=O)CC[C@@]36O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C20H23NO4/c22-13-4-3-12-9-15-20(24)6-5-14(23)18-19(20,16(12)17(13)25-18)7-8-21(15)10-11-1-2-11/h3-4,11,15,18,22,24H,1-2,5-10H2/t15-,18+,19+,20-/m1/s1
Formula Weight 341.1627
Chemical Name or Material Naltrexone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.