missing translation for 'onlineSavingsMsg'
Learn More

N-Formyl Benzocaine, TRC

Product Code. 30010520
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010520 100 mg 100mg
30137277 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30010520 Supplier Toronto Research Chemicals Supplier No. TRCF697300100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5422-63-9
Molecular Formula C10 H11 N O3
Molecular Weight (g/mol) 193.199
Synonym 4-(Formylamino)benzoic Acid Ethyl Ester, Benzoic acid, p-formamido-, ethyl ester (6CI), Ethyl 4-(formylamino)benzoate, Ethyl 4-formamidobenzoate, Ethyl p-formamidobenzoate, NSC 12009, Ethyl 4-Formamidobenzoate, N-Formylbenzocaine, N-Formyl benzocaine
IUPAC Name ethyl 4-formamidobenzoate
SMILES CCOC(=O)c1ccc(NC=O)cc1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H11NO3/c1-2-14-10(13)8-3-5-9(6-4-8)11-7-12/h3-7H,2H2,1H3,(H,11,12)
Formula Weight 193.074
Percent Purity >95%
Chemical Name or Material Ethyl 4-Formamidobenzoate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.