missing translation for 'onlineSavingsMsg'
Learn More

Methyl Acetylsalicylate, Mikromol™

Product Code. 30050263
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 30050263

Brand: Mikromol MM0133.12

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Methyl Acetylsalicylate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl Acetylsalicylate
CAS 580-02-9
Analyte or Component Names Methyl Acetylsalicylate
Shipping Condition Room Temperature
Molecular Formula C10 H10 O4
Certifications/Compliance ISO 17025
Synonym Methyl Acetylsalicylate, 2-(Acetyloxy)-benzoic acid methyl ester, Acetylsalicylic acid, methyl ester (6CI), Salicylic acid acetate, methyl ester (8CI), Salicylic acid, methyl ester, acetate (7CI), Aspirin methyl ester, Methyl 2-acetoxybenzoate, Methyl O-acetylsalicylate, Methyl acetylsalicylate, Methyl aspirin, Methyl o-acetoxybenzoate, Methyl rhodin, Methylrhodine, NSC 403847
InChI Formula InChI=1S/C10H10O4/c1-7(11)14-9-6-4-3-5-8(9)10(12)13-2/h3-6H,1-2H3
SMILES COC(=O)c1ccccc1OC(=O)C
IUPAC Name methyl 2-acetyloxybenzoate
Molecular Weight (g/mol) 194.18
Quantity 100 mg
Formula Weight 194.0579 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.