missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mercuric Trifluoroacetate, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Mercuric Trifluoroacetate |
| CAS | 13257-51-7 |
| Molecular Formula | 2(C2HF3O2) . Hg |
| Quantity | 100 mg |
| Synonym | Bis(trifluoroacetato)mercury, Bis(trifluoroacetoxy)mercury, Mercury Bis(trifluoroacetate), Mercury(2+) Trifluoroacetate, Mercury(II) Bis(trifluoroacetate), Mercury(II) Trifluoroacetate, Trifluoroacetic Acid Mercury(2+) Salt |
| InChI Formula | InChI=1S/2C2HF3O2.Hg/c2*3-2(4,5)1(6)7;/h2*(H,6,7); |
| SMILES | [Hg].OC(=O)C(F)(F)F.OC(=O)C(F)(F)F |
| IUPAC Name | mercury;2,2,2-trifluoroacetic acid |
| Molecular Weight (g/mol) | 426.62 |
| Formula Weight | 429.9564 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction