missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iopamidol, MedChemExpress
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Description
Iopamidol is a nonionic, X-Ray iodinated contrast agent (CA) for a wide variety of diagnostic applications. Iopamidol contains amide and hydroxyl functionalities that can be exploited for the generation of the chemical exchange saturation transfer (CEST) contrast.
Specifications
Specifications
| Chemical Name or Material | Iopamidol |
| CAS | 60166-93-0 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C17H22I3N3O8 |
| Quantity | 100 mg |
| Synonym | B-15000 SQ-13396 |
| Solubility Information | H2O : 100 mg/mL (128.69 mM; Need ultrasonic) ∣DMSO : 50 mg/mL (64.34 mM; Need ultrasonic) |
| SMILES | O=C(C1=C(I)C(NC([C@@H](O)C)=O)=C(I)C(C(NC(CO)CO)=O)=C1I)NC(CO)CO |
| Molecular Weight (g/mol) | 777.09 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction