missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Imipramine.HCl, Lipomed
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Imipramine.HCl
Specifications
Specifications
| Chemical Name or Material | Imipramine hydrochloride |
| CAS | 113-52-0 |
| Analyte or Component Names | Imipramine hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H24 N2 . Cl H |
| Synonym | Clomipramine Hydrochloride Imp. B (EP), Clomipramine Imp. B (EP), Trimipramine Maleate Imp. D (EP), Trimipramine Imp. D (EP), Imipramine Hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | Cl.CN(C)CCCN1c2ccccc2CCc3ccccc13 |
| IUPAC Name | 3-(5,6-dihydrobenzo[b][1 ]benzazepin-11-yl)-N,N-dimethylpropan-1-amine;hydrochloride |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction