missing translation for 'onlineSavingsMsg'
Learn More

Hesperadin, MedChemExpress

Product Code. 16405736
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16405736

Brand: Medchem Express HY12054

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Hesperadin is an ATP competitive indolinone inhibitor of Aurora A and B. Hesperadin inhibits Aurora B with an IC50 of 250 nM. Hesperadin inhibits the growth of Trypanosoma brucei by blocking nuclear division and cytokinesis. Hesperadin also is a broad-spectrum influenza antiviral.

Hesperadin is an ATP competitive indolinone inhibitor of Aurora A and B. Hesperadin inhibits Aurora B with an IC50 of 250 nM. Hesperadin inhibits the growth of Trypanosoma brucei by blocking nuclear division and cytokinesis. Hesperadin also is a broad-spectrum influenza antiviral.

Specifications

Chemical Name or Material Hesperadin
Physical Form Solid
CAS 422513-13-1
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C29H32N4O3S
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : ≥ 100 mg/mL (193.55 mM)
SMILES O=C1NC2=CC=C(C=C2/C1=C(NC3=CC=C(C=C3)CN4CCCCC4)\C5=CC=CC=C5)NS(CC)(=O)=O
Molecular Weight (g/mol) 516.65
Formula Weight 516.65
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.