missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Halofantrine Hydrochloride, Mikromol™
Description
Halofantrine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Halofantrine hydrochloride |
| CAS | 36167-63-2 |
| Analyte or Component Names | Halofantrine hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C26 H30 Cl2 F3 N O . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Halofantrine Hydrochloride, 9-[3-(Dibutylamino)-1-hydroxypropyl]-1,3-dichloro-6-(trifluoromethyl)phenanthrene hydrochloride, Halofantrine hydrochloride, SKF 102886, WR 171669, 1,3-Dichloro-alpha-[2-(dibutylamino)ethyl]-6-(trifluoromethyl)-9-phenanthrenemethanol hydrochloride, (+/-)-Halofantrine hydrochloride, SKF-102886-A |
| InChI Formula | InChI=1S/C26H30Cl2F3NO.ClH/c1-3-5-10-32(11-6-4-2)12-9-25(33)23-16-22-21(14-18(27)15-24(22)28)20-13-17(26(29,30)31)7-8-19(20)23;/h7-8,13-16,25,33H,3-6,9-12H2,1-2H3;1H |
| SMILES | Cl.CCCCN(CCCC)CCC(O)c1cc2c(Cl)cc(Cl)cc2c3cc(ccc13)C(F)(F)F |
| IUPAC Name | 3-(dibutylamino)-1-[1,3-dichloro-6-(trifluoromethyl)phenanthren-9-yl]propan-1-ol;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?