missing translation for 'onlineSavingsMsg'
Learn More

H-Val-Gln-OH, TRC

Product Code. 30451795
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30451795

Brand: Toronto Research Chemicals TRCV191745100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material H-Val-Gln-OH
CAS 42854-54-6
Molecular Formula C10 H19 N3 O4
Quantity 100 mg
Synonym L-Glutamine','L-valyl- (9CI','ACI), L-Glutamine','N2-L-valyl- (ZCI), L-Valyl-L-glutamine (ACI), Glutamine','N2-L-valyl- (7CI), 113: PN: WO2011146121 PAGE: 116 claimed sequence, 27: PN: EP2161028 PAGE: 10 claimed protein, 471: PN: WO2005081628 SEQID: 1580 claimed protein, 8: PN: JP2007217358 SEQID: 8 claimed protein, 98: PN: US20070066537 PAGE: 17 claimed protein
InChI Formula InChI=1S/C10H19N3O4/c1-5(2)8(12)9(15)13-6(10(16)17)3-4-7(11)14/h5-6,8H,3-4,12H2,1-2H3,(H2,11,14)(H,13,15)(H,16,17)/t6-,8-/m0/s1
SMILES CC(C)[C@H](N)C(=O)N[C@@H](CCC(=O)N)C(=O)O
IUPAC Name (2S)-5-amino-2-[[(2S)-2-amino-3-methyl-butanoyl]amino]-5-oxo-pentanoic acid
Molecular Weight (g/mol) 245.276
Formula Weight 245.1376
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.