missing translation for 'onlineSavingsMsg'
Learn More

Flurbiprofen Ethyl Ester, Mikromol™

Product Code. 30051415
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30051415

Brand: Mikromol MM0727.10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Flurbiprofen Ethyl Ester
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Flurbiprofen Ethyl Ester
CAS 64858-90-8
Analyte or Component Names Flurbiprofen Ethyl Ester
Shipping Condition Room Temperature
Molecular Formula C17 H17 F O2
Certifications/Compliance ISO 9001
Synonym Flurbiprofen Ethyl Ester, 2-Fluoro-alpha-methyl-[1,1'-biphenyl]-4-acetic acid ethyl ester, Flurbiprofen ethyl ester
InChI Formula InChI=1S/C17H17FO2/c1-3-20-17(19)12(2)14-9-10-15(16(18)11-14)13-7-5-4-6-8-13/h4-12H,3H2,1-2H3
SMILES CCOC(=O)C(C)c1ccc(c(F)c1)c2ccccc2
IUPAC Name ethyl 2-(3-fluoro-4-phenylphenyl)propanoate
Molecular Weight (g/mol) 272.31
Quantity 100 mg
Formula Weight 272.1213 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.