missing translation for 'onlineSavingsMsg'
Learn More

Etofenamate, MedChemExpress

Product Code. 16447256
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 16447256

Brand: Medchem Express HY17361

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Etofenamate, a non-steroid anti-inflammatory drug (NSAID) and a non-selective COX inhibitor, possesses analgesic, anti-rheumatic, antipyretic and anti-inflammatory properties. Etofenamate is used in the research for osteoarthritis, arthritis and other inflammatory diseases.

Etofenamate, a non-steroid anti-inflammatory drug (NSAID) and a non-selective COX inhibitor, possesses analgesic, anti-rheumatic, antipyretic and anti-inflammatory properties. Etofenamate is used in the research for osteoarthritis, arthritis and other inflammatory diseases.

Specifications

Chemical Name or Material Etofenamate
Physical Form Oil
CAS 30544-47-9
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C18H18F3NO4
For Use With (Application) COVID-19-immunoregulation
Solubility Information Ethanol : 120 mg/mL (324.90 mM; Need ultrasonic) ∣DMSO : ≥ 50 mg/mL (135.38 mM)
SMILES O=C(OCCOCCO)C1=CC=CC=C1NC2=CC=CC(C(F)(F)F)=C2
Molecular Weight (g/mol) 369.34
Formula Weight 369.34
Percent Purity 98.14%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.