missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-(2-Oxopyrrolidin-1-yl)acetate (80%), TRC

Product Code. 30011232
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011232 100 mg 100mg
30135635 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011232 Supplier Toronto Research Chemicals Supplier No. TRCE925885100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61516-73-2
Molecular Formula C8 H13 N O3
Molecular Weight (g/mol) 171.19
Synonym Ethyl (2-Oxopyrrolidin-1-yl)acetate, Piracetam Imp. C (EP), 2-Oxo-1-pyrrolidineacetic acid ethyl ester, 1-(Ethoxycarbonylmethyl)-2-pyrrolidone, Ethyl (2-oxo-1-pyrrolidinyl)acetate, Ethyl 2-oxo-1-pyrrolidineacetate
IUPAC Name ethyl 2-(2-oxopyrrolidin-1-yl)acetate
SMILES CCOC(=O)CN1CCCC1=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H13NO3/c1-2-12-8(11)6-9-5-3-4-7(9)10/h2-6H2,1H3
Formula Weight 171.0895
Chemical Name or Material Ethyl (2-Oxopyrrolidin-1-yl)acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.