missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Estra-1,3,5(10)-triene-3,16beta,17beta-triol (16-epi-Estriol), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Estra-1,3,5(10)-triene-3,16beta,17beta-triol (16-epi-Estriol)
Specifications
Specifications
| Chemical Name or Material | 16-Epiestriol |
| CAS | 547-81-9 |
| Analyte or Component Names | 16-Epiestriol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H24 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Estra-1,3,5(10)-triene-3,16beta,17beta-triol, Estriol Imp. F (EP), 16-epi-Estriol, 16-Epiestratriol, 16-Epiestriol, Actriol, Epiestriol, Epioestriol, NSC 26646, Estra-1,3,5(10)-triene-3,16,17-triol, (16beta,17beta)-, Estra-1,3,5(10)-triene-3,16beta,17-triol (7CI), Estra-1,3,5(10)-triene-3,16beta,17beta-triol (8CI), (16beta,17beta)-Estra-1,3,5(10)-triene-3,16,17-triol, 16beta,17beta-Estriol, 16beta-Hydroxyestradiol, Estriol Impurity F |
| InChI Formula | InChI=1S/C18H24O3/c1-18-7-6-13-12-5-3-11(19)8-10(12)2-4-14(13)15(18)9-16(20)17(18)21/h3,5,8,13-17,19-21H,2,4,6-7,9H2,1H3/t13-,14-,15+,16+,17+,18+/m1/s1 |
| SMILES | C[C@]12CC[C@H]3[C@@H](CCc4cc(O)ccc34)[C@@H]1C[C@H](O)[C@@H]2O |
| IUPAC Name | (8R,9S,13S,14S,16S,17R)-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,16,17-triol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction