missing translation for 'onlineSavingsMsg'
Learn More

Dimethadione, Mikromol™

Product Code. 30051108
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30051108

Brand: Mikromol MM3048.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Dimethadione
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimethadione
CAS 695-53-4
Analyte or Component Names Dimethadione
Shipping Condition Room Temperature
Molecular Formula C5 H7 N O3
Certifications/Compliance ISO 17025
Synonym 2,4-Oxazolidinedione, 5,5-dimethyl-, 5,5-Dimethyl-1,3-oxazolidine-2,4-dione, 5,5-Dimethyl-2,4-oxazolidinedione, 5,5-Dimethyloxazolidinedione, AC 1198, BAX 1400Z, BAY 1400Z, DMO, Dimethadion, Dimethadione, Eupractone, NSC 30152, Nortrimethadione
InChI Formula InChI=1S/C5H7NO3/c1-5(2)3(7)6-4(8)9-5/h1-2H3,(H,6,7,8)
SMILES CC1(C)OC(=O)NC1=O
IUPAC Name 5,5-dimethyl-1,3-oxazolidine-2,4-dione
Molecular Weight (g/mol) 129.11
Quantity 100 mg
Formula Weight 129.0426 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.