missing translation for 'onlineSavingsMsg'
Learn More

Diclofenac 2,3-Butylene Glycol Ester, Mikromol™

Product Code. 30050901
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30050901

Brand: Mikromol MM0006.38

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Diclofenac 2,3-Butylene Glycol Ester
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diclofenac 2,3-Butylene Glycol Ester
CAS 95093-57-5
Analyte or Component Names Diclofenac 2,3-Butylene Glycol Ester
Shipping Condition Room Temperature
Molecular Formula C18 H19 Cl2 N O3
Certifications/Compliance ISO 9001
Synonym Diclofenac 2,3-Butylene Glycol Ester, Benzeneacetic acid, 2-[(2,6-dichlorophenyl)amino]-, 2-hydroxy-1-methylpropyl ester
InChI Formula InChI=1S/C18H19Cl2NO3/c1-11(22)12(2)24-17(23)10-13-6-3-4-9-16(13)21-18-14(19)7-5-8-15(18)20/h3-9,11-12,21-22H,10H2,1-2H3
SMILES CC(O)C(C)OC(=O)Cc1ccccc1Nc2c(Cl)cccc2Cl
IUPAC Name 3-hydroxybutan-2-yl 2-[2-(2,6-dichloroanilino)phenyl]acetate
Molecular Weight (g/mol) 368.25
Quantity 100 mg
Formula Weight 367.0742 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.