missing translation for 'onlineSavingsMsg'
Learn More

Diazoxide, Mikromol™

Product Code. 17923096
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 17923096

Brand: Mikromol MM3038.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Diazoxide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diazoxide
CAS 364-98-7
Analyte or Component Names Diazoxide
Shipping Condition Room Temperature
Molecular Formula C8 H7 Cl N2 O2 S
Certifications/Compliance ISO 17034
Synonym Diazoxide, 7-chloro-3-methyl-2H-1,2,4-benzothiadiazine 1,1-dioxide, 2H-1,2,4-Benzothiadiazine, 7-chloro-3-methyl-, 1,1-dioxide, 4H-1,2,4-Benzothiadiazine, 7-chloro-3-methyl-, 1,1-dioxide (7CI), 3-Methyl-7-chloro-1,2,4-benzothiadiazine 1,1-dioxide, 7-Chloro-3-methyl-2H-1,2,4-benzothiadiazine 1,1-dioxide, Diazoxide, Dizoxide, Eudemine injection, Hyperstat, Hypertonalum, Mutabase, NSC 64198, NSC 76130, Proglicem, Proglycem, SRG 95213, Sch 6783
InChI Formula InChI=1S/C8H7ClN2O2S/c1-5-10-7-3-2-6(9)4-8(7)14(12,13)11-5/h2-4H,1H3,(H,10,11)
SMILES CC1=Nc2ccc(Cl)cc2S(=O)(=O)N1
IUPAC Name 7-chloro-3-methyl-2H-1λ^{6},2,4-benzothiadiazine 1,1-dioxide
Molecular Weight (g/mol) 230.67
Quantity 100 mg
Formula Weight 229.9917 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.