missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dexibuprofen 2,3-Butylene Glycol Ester, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Dexibuprofen 2,3-Butylene Glycol Ester
Specifications
Specifications
| Chemical Name or Material | Dexibuprofen 2,3-Butylene Glycol Ester |
| CAS | 2983462-55-9 |
| Analyte or Component Names | Dexibuprofen 2,3-Butylene Glycol Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H26 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Dexibuprofen 2,3-Butylene Glycol Ester |
| InChI Formula | InChI=1S/C17H26O3/c1-11(2)10-15-6-8-16(9-7-15)12(3)17(19)20-14(5)13(4)18/h6-9,11-14,18H,10H2,1-5H3/t12-,13?,14?/m0/s1 |
| SMILES | CC(C)Cc1ccc(cc1)[C@H](C)C(=O)OC(C)C(C)O |
| IUPAC Name | 3-hydroxybutan-2-yl (2S)-2-[4-(2-methylpropyl)phenyl]propanoate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction