missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bupropion.HCl, Lipomed
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Bupropion.HCl
Specifications
Specifications
| Chemical Name or Material | Bupropion (hydrochloride) |
| CAS | 31677-93-7 |
| Analyte or Component Names | Bupropion (hydrochloride) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H18 Cl N O . Cl H |
| Synonym | 1-Propanone, 1-(3-chlorophenyl)-2-[(1,1-dimethylethyl)amino]-, hydrochloride (1:1), 1-Propanone, 1-(3-chlorophenyl)-2-[(1,1-dimethylethyl)amino]-, hydrochloride (9 CI), Bupropion hydrochloride, NSC 315851, Wellbatrin, GI 267196 X, GI 267196 X, GI 267196 X, GI 267196 X, GI 267196 X, GI 267196 X, GI 267196 X, GI 267196 X, GI 267196 X, GI 267196 X, GI 267196 X, GI 267196 X, GI 267196 X |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | Cl.CC(NC(C)(C)C)C(=O)c1cccc(Cl)c1 |
| IUPAC Name | 2-(tert-butylamino)-1-(3-chlorophenyl)propan-1-one;hydrochloride |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction