missing translation for 'onlineSavingsMsg'
Learn More

Biprofen, TRC

Product Code. 30011769
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011769 100 mg 100mg
30136246 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011769 Supplier Toronto Research Chemicals Supplier No. TRCB399050100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6341-72-6
Molecular Formula C15 H14 O2
Molecular Weight (g/mol) 226.27
IUPAC Name 2-(4-phenylphenyl)propanoic acid
SMILES CC(C(=O)O)c1ccc(cc1)c2ccccc2

Specifications

Quantity 100 mg
Synonym (2RS)-2-(Biphenyl-4-yl)propanoic Acid, Flurbiprofen Imp. A (EP), Flurbiprofen Related Compound A (USP), 2-(Biphenyl-4-yl)propionic acid, Biprofen, [1,1'-Biphenyl]-4-acetic acid, α-methyl-, 4-Biphenylacetic acid, α-methyl- (8CI), Hydratropic acid, p-phenyl- (6CI,7CI), α-Methyl[1,1'-biphenyl]-4-acetic acid, (±)-2-(4-Biphenyl)propionic acid, 16300Y1, 2-((1,1'-Biphenyl]-4-yl)propanoic acid, 2-(4-Biphenyl)propionic acid, 2-(4-Biphenylyl)propanoic acid, 2-(4-Biphenylyl)propionic acid, 2-(4-Phenylphenyl)propionic acid, NSC 16300
InChI Formula InChI=1S/C15H14O2/c1-11(15(16)17)12-7-9-14(10-8-12)13-5-3-2-4-6-13/h2-11H,1H3,(H,16,17)
Formula Weight 226.0994
Percent Purity >95%
Chemical Name or Material (2RS)-2-(biphenyl-4-yl)propanoic acid
Grade HPLC
compliance-icons
Product Identifier
  • Biprofen
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P330-Rinse mouth.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contain: nyl]-4-acetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.