missing translation for 'onlineSavingsMsg'
Learn More

Bedaquiline fumarate, MedChemExpress

Product Code. 16436586
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16436586

Brand: Medchem Express HY14881A100mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bedaquiline fumarate, a diarylquinoline antibiotic that targets ATP synthase, is effective for the treatment of Mycobacterium tuberculosis infections.

Bedaquiline fumarate, a diarylquinoline antibiotic that targets ATP synthase, is effective for the treatment of Mycobacterium tuberculosis infections.

Specifications

Chemical Name or Material Bedaquiline fumarate
Physical Form Solid
CAS 845533-86-0
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C36H35BrN2O6
For Use With (Application) COVID-19-immunoregulation
Synonym R403323 TMC207 fumarate R207910 fumarate
Solubility Information DMSO : 100 mg/mL (148.90 mM; Need ultrasonic)
SMILES OC(/C=C/C(O)=O)=O.BrC1=CC=C(N=C(OC)C([C@H]([C@@](C2=CC=CC3=C2C=CC=C3)(O)CCN(C)C)C4=CC=CC=C4)=C5)C5=C1
Molecular Weight (g/mol) 671.58
Formula Weight 671.58
Percent Purity 99.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.