missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5'-Xanthylic Acid Triethylammonium Salt, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | 5'-Xanthylic Acid Triethylammonium Salt |
| Molecular Formula | C10 H13 N4 O9 P . C6 H15 N |
| Quantity | 100 mg |
| Synonym | Xanthosine 5’-Monophosphate Triethylammonium Salt |
| InChI Formula | InChI=1S/C10H13N4O9P.C6H15N/c15-5-3(1-22-24(19,20)21)23-9(6(5)16)14-2-11-4-7(14)12-10(18)13-8(4)17;1-4-7(5-2)6-3/h2-3,5-6,9,15-16H,1H2,(H2,19,20,21)(H2,12,13,17,18);4-6H2,1-3H3/t3-,5?,6?,9-;/m1./s1 |
| SMILES | CCN(CC)CC.OC1C(O)[C@@H](O[C@@H]1COP(=O)(O)O)n2cnc3C(=O)NC(=O)Nc23 |
| IUPAC Name | N,N-diethylethanamine;[(2R,5R)-5-(2,6-dioxo-3H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate |
| Molecular Weight (g/mol) | 465.395 |
| Formula Weight | 465.162 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction