missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxy-1H-benzimidazole-2-thiol, Mikromol™

Product Code. 17931246
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 17931246

Brand: Mikromol MM0095.03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

5-Methoxy-1H-benzimidazole-2-thiol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Methoxy-1H-benzimidazole-2-thiol
CAS 37052-78-1
Analyte or Component Names 5-Methoxy-1H-benzimidazole-2-thiol
Shipping Condition Room Temperature
Molecular Formula C8 H8 N2 O S
Certifications/Compliance ISO 17025
Synonym Esomeprazole Magnesium Trihydrate Imp. A (EP), Esomeprazole Imp. A (EP), Omeprazole Imp. A (EP), Omeprazole USP Related Compound B, Omeprazole USP RC B, 5-Methoxy-1H-benzimidazole-2-thiol, Omeprazole Related Compound B, Esomeprazole Magnesium Trihydrate Impurity A, Esomeprazole Sodium Impurity A, Omeprazole Impurity A, Esomeprazole Impurity A, Esomeprazole Impurity A
InChI Formula InChI=1S/C8H8N2OS/c1-11-5-2-3-6-7(4-5)10-8(12)9-6/h2-4H,1H3,(H2,9,10,12)
SMILES COc1ccc2[nH]c(S)nc2c1
IUPAC Name 5-methoxy-1H-benzimidazole-2-thiol
Molecular Weight (g/mol) 180.23
Quantity 100 mg
Formula Weight 180.0357 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.