missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxytryptophan, MedChemExpress

Product Code. 16408996
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16408996

Brand: Medchem Express HYN0122

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 5-Hydroxytryptophan, a tryptophan metabolite, is a direct 5-hydroxytryptamine (5-HT) precursor and an L-aromatic amino acid decarboxylase substrate. .

5-Hydroxytryptophan, a tryptophan metabolite, is a direct 5-hydroxytryptamine (5-HT) precursor and an L-aromatic amino acid decarboxylase substrate. .

Specifications

Chemical Name or Material 5-Hydroxytryptophan
CAS 56-69-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C11H12N2O3
Synonym 5-HTP DL-5-Hydroxytryptophan
Solubility Information H2O : 12.5 mg/mL (56.76 mM; Need ultrasonic)
SMILES NC(CC1=CNC2=CC=C(O)C=C12)C(O)=O
Molecular Weight (g/mol) 220.22
Quantity 100 mg
Formula Weight 220.22
Percent Purity 98.99%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.