missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Baclofen EP Impurity A, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Baclofen EP Impurity A
Specifications
Specifications
| Chemical Name or Material | (4RS)-4-(4-Chlorophenyl)pyrrolidin-2-one |
| CAS | 22518-27-0 |
| Analyte or Component Names | (4RS)-4-(4-Chlorophenyl)pyrrolidin-2-one |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H10 Cl N O |
| Certifications/Compliance | ISO 17025 |
| Synonym | (4RS)-4-(4-Chlorophenyl)pyrrolidin-2-one (Baclofen Lactam), (RS)-4-amino-3-(4-chlorophenyl)butyric acid lactam, 4-(p-Chlorophenyl)-2-pyrrolidinone, 4-(4-Chlorophenyl)-2-pyrrolidinone, 4-(4-Chlorophenyl)-2-pyrrolidone, Baclofen Related Compound A, Baclofen Imp. A (EP), (+/-)-Baclofen lactam, Baclofen Related Compound A, Baclofen Impurity A |
| InChI Formula | InChI=1S/C10H10ClNO/c11-9-3-1-7(2-4-9)8-5-10(13)12-6-8/h1-4,8H,5-6H2,(H,12,13) |
| SMILES | Clc1ccc(cc1)C2CNC(=O)C2 |
| IUPAC Name | 4-(4-chlorophenyl)pyrrolidin-2-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction