missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(3RS)-6-Cyano-3-[3-(dimethylamino)propyl]-3-(4-fluorophenyl)isobenzofuran-1(3H)-one, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
(3RS)-6-Cyano-3-[3-(dimethylamino)propyl]-3-(4-fluorophenyl)isobenzofuran-1(3H)-one
Specifications
Specifications
| Chemical Name or Material | (3RS)-6-Cyano-3-[3-(dimethylamino)propyl]-3-(4-fluorophenyl)isobenzofuran-1(3H)-one |
| CAS | 372941-54-3 |
| Analyte or Component Names | (3RS)-6-Cyano-3-[3-(dimethylamino)propyl]-3-(4-fluorophenyl)isobenzofuran-1(3H)-one |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H19 F N2 O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Citalopram Hydrobromide Imp. C (EP), Citalopram Imp. C (EP), Citalopram USP Related Compound C, Citalopram USP RC C, 5-Isobenzofurancarbonitrile, 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-3-oxo-, 1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-3-oxo-5-isobenzofurancarbonitrile, 3-Oxo Citalopram, (3RS)-6-Cyano-3-[3-(dimethylamino)propyl]-3-(4-fluorophenyl)isobenzofuran-1(3H)-one, Citalopram Related Compound C, Citalopram Hydrobromide Impurity C, Escitalopram Oxalate Impurity C, Citalopram Impurity C, Escitalopram Impurity C |
| InChI Formula | InChI=1S/C20H19FN2O2/c1-23(2)11-3-10-20(15-5-7-16(21)8-6-15)18-9-4-14(13-22)12-17(18)19(24)25-20/h4-9,12H,3,10-11H2,1-2H3 |
| SMILES | CN(C)CCCC1(OC(=O)c2cc(ccc12)C#N)c3ccc(F)cc3 |
| IUPAC Name | 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-3-oxo-2-benzofuran-5-carbonitrile |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction