missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[(RS)-[(3,5-Dimethylpyridin-2-yl)methyl]sulphinyl]-5-methoxy-1H-benzimidazole, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
2-[(RS)-[(3,5-Dimethylpyridin-2-yl)methyl]sulphinyl]-5-methoxy-1H-benzimidazole
Specifications
Specifications
| Chemical Name or Material | 2-[(RS)-[(3,5-Dimethylpyridin-2-yl)methyl]sulphinyl]-5-methoxy-1H-benzimidazole |
| CAS | 110374-16-8 |
| Analyte or Component Names | 2-[(RS)-[(3,5-Dimethylpyridin-2-yl)methyl]sulphinyl]-5-methoxy-1H-benzimidazole |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C16 H17 N3 O2 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | Esomeprazole Magnesium Trihydrate Imp. B (EP), Esomeprazole Imp. B (EP), Omeprazole Imp. B (EP), 2-[(RS)-[(3,5-Dimethylpyridin-2-yl)methyl]sulphinyl]-5-methoxy-1H-benzimidazole, Esomeprazole Magnesium Trihydrate Impurity B, Omeprazole Impurity B, Esomeprazole Impurity B |
| InChI Formula | InChI=1S/C16H17N3O2S/c1-10-6-11(2)15(17-8-10)9-22(20)16-18-13-5-4-12(21-3)7-14(13)19-16/h4-8H,9H2,1-3H3,(H,18,19) |
| SMILES | COc1ccc2[nH]c(nc2c1)S(=O)Cc3ncc(C)cc3C |
| IUPAC Name | 2-[(3,5-dimethylpyridin-2-yl)methylsulfinyl]-5-methoxy-1H-benzimidazole |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction