missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Methylethenyl)-6-(1-methylethyl)phenol, Mikromol™

Product Code. 30049421
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30049421

Brand: Mikromol MM0461.10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-(1-Methylethenyl)-6-(1-methylethyl)phenol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(1-Methylethenyl)-6-(1-methylethyl)phenol
CAS 74926-89-9
Analyte or Component Names 2-(1-Methylethenyl)-6-(1-methylethyl)phenol
Shipping Condition Room Temperature
Molecular Formula C12H16O
Certifications/Compliance ISO 9001
Synonym 2-(1-Methylethenyl)-6-(1-methylethyl)phenol, Propofol Imp. B (EP), Propofol Impurity B
InChI Formula InChI=1S/C12H16O/c1-8(2)10-6-5-7-11(9(3)4)12(10)13/h5-7,9,13H,1H2,2-4H3
SMILES CC(C)c1cccc(C(=C)C)c1O
IUPAC Name 2-isopropenyl-6-isopropyl-phenol
Molecular Weight (g/mol) 176.25
Quantity 100 mg
Formula Weight 176.1201 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.