missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tool Steel, AISI T1, T12001, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Tool Steel, AISI T1, T12001, 100g
Specifications
Specifications
| Chemical Name or Material | Titanium/Tin/Nitrogen/Phosphorus/Copper/Nickel/Silicon/Manganese/Cobalt/Molybdenum/Carbon/Vanadium/Chromium/Tungsten/Calcium/Boron/Niobium/Zirconium/Oxygen/Aluminium/Arsenic |
| CAS | 7439-89-6 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Titanium 0.004 %/Tin 0.019 %/Nitrogen 0.0221 %/Phosphorus 0.036 %/Copper 0.13 %/Nickel 0.23 %/Silicon 0.29 %/Manganese 0.33 %/Cobalt 0.39 %/Molybdenum 0.4 %/Carbon 0.79 %/Vanadium 1.13 %/Chromium 4.32 %/Tungsten 17.99 %/Calcium (0.0006) %/Boron (0.0018) %/Niobium (0.003) %/Zirconium (0.006) %/Oxygen (0.0107) %/Aluminium (0.016) %/Arsenic (0.051) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction