missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, AISI 410, S41000, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Stainless Steel, AISI 410, S41000, 100g
Specifications
Specifications
| Chemical Name or Material | Boron/Titanium/Niobium/Aluminium/Arsenic/Tantalum/Tungsten/Oxygen/Tin/Cobalt/Vanadium/Nitrogen/Phosphorus/Molybdenum/Copper/Carbon/Nickel/Silicon/Manganese/Chromium |
| CAS | 7439-89-6 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Boron 0.0005 %/Titanium 0.0016 %/Niobium 0.0023 %/Aluminium 0.003 %/Arsenic 0.003 %/Tantalum 0.004 %/Tungsten 0.004 %/Oxygen 0.0053 %/Tin 0.006 %/Cobalt 0.0213 %/Vanadium 0.022 %/Nitrogen 0.0225 %/Phosphorus 0.033 %/Molybdenum 0.038 %/Copper 0.11 %/Carbon 0.143 %/Nickel 0.2 %/Silicon 0.38 %/Manganese 0.57 %/Chromium 12.51 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction