missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, AISI 316H, S31609, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Stainless Steel, AISI 316H, S31609, 100g
Specifications
Specifications
| Chemical Name or Material | Boron/Calcium/Tin/Titanium/Zirconium/Aluminium/Phosphorus/Niobium/Nitrogen/Cobalt/Vanadium/Tungsten/Oxygen/Copper/Carbon/Silicon/Manganese/Molybdenum/Nickel/Chromium/Magnesium/Arsenic/Zinc/Lead/Tantalum |
| CAS | 7439-89-6 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Boron 0.0006 %/Calcium 0.0014 %/Tin 0.002 %/Titanium 0.002 %/Zirconium 0.003 %/Aluminium 0.004 %/Phosphorus 0.004 %/Niobium 0.005 %/Nitrogen 0.006 %/Cobalt 0.007 %/Vanadium 0.007 %/Tungsten 0.0119 %/Oxygen 0.016 %/Copper 0.021 %/Carbon 0.16 %/Silicon 0.64 %/Manganese 1.71 %/Molybdenum 2.79 %/Nickel 12.9 %/Chromium 17 %/Magnesium (0.0001) %/Arsenic (0.001) %/Zinc (0.001) %/Lead (0.002) %/Tantalum (0.005) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction