Learn More
IRIS Biotech 100GR Fmoc-Rink-Amid MBHA resin > 0.4 mmol/g 1% DVB
Brand:  IRIS Biotech BR-1300.0100
Encompass
100GR Fmoc-Rink-Amid MBHA resin > 0.4 mmol/g 1% DV
Product Code. 17260973
  • £1310.55 / 100g