missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferromanganese Powder, ARMI∣MBH
Description
Ferromanganese Powder, 100 grams
Specifications
Specifications
| Chemical Name or Material | Boron/Sulfur/Copper/Nickel/Cobalt/Chromium/Phosphorus/Nitrogen/Silicon/Carbon/Iron/Manganese |
| CAS | 7439-89-6 |
| Quantity | 100 g |
| Form | Fine Powder |
| Concentration or Composition (by Analyte or Components) | Boron 0.0019 %/Sulfur 0.008 %/Copper 0.01 %/Nickel 0.015 %/Cobalt 0.019 %/Chromium 0.036 %/Phosphorus 0.08 %/Nitrogen 0.083 %/Silicon 0.42 %/Carbon 1.13 %/Iron 16.6 %/Manganese 79.7 % |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?