missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Lead-Free Brass, CDA 693, C69300, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Copper, Lead-Free Brass, CDA 693, C69300, 100g
Specifications
Specifications
| Chemical Name or Material | Boron/Arsenic/Silver/Nickel/Tin/Iron/Antimony/Lead/Phosphorus/Silicon/Zinc/Copper/Nitrogen/Beryllium/Cadmium/Magnesium/Zirconium/Aluminium/Bismuth/Chromium/Manganese/Niobium/Oxygen/Selenium/Carbon/Tellurium/Cobalt |
| CAS | 7440-50-8 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Boron 0.0008 %/Arsenic 0.001 %/Silver 0.0017 %/Nickel 0.002 %/Tin 0.006 %/Iron 0.011 %/Antimony 0.014 %/Lead 0.042 %/Phosphorus 0.09 %/Silicon 3.09 %/Zinc 21.4 %/Copper 75.4 %/Nitrogen (0.0001) %/Beryllium (0.0003) %/Cadmium (0.0003) %/Magnesium (0.0003) %/Zirconium (0.0004) %/Aluminium (0.001) %/Bismuth (0.001) %/Chromium (0.001) %/Manganese (0.001) %/Niobium (0.001) %/Oxygen (0.001) %/Selenium (0.001) %/Carbon (0.002) %/Tellurium (0.003) %/Cobalt (0.004) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction