missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Silicon Bronze, CDA 647, C64700, 100g, ARMI∣MBH
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
missing translation for 'unitSize'
100g
Description
Copper, Silicon Bronze, CDA 647, C64700, 100g
Specifications
Specifications
| Chemical Name or Material | Aluminium/Zinc/Iron/Magnesium/Zirconium/Manganese/Silicon/Nickel/Hydrogen/Tungsten/Vanadium/Cobalt/Arsenic/Bismuth/Chromium/Molybdenum/Titanium/Silver/Lead/Selenium/Tin/Antimony/Phosphorus/Cadmium/Nitrogen/Oxygen/Sulfur/Niobium/Carbon/Boron/Copper |
| CAS | 7440-50-8 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Aluminium 0.0027 %/Zinc 0.005 %/Iron 0.0056 %/Magnesium 0.0086 %/Zirconium 0.044 %/Manganese 0.099 %/Silicon 0.7 %/Nickel 2.69 %/Hydrogen (<0.0010) %/Tungsten (<0.0010) %/Vanadium (<0.0010) %/Cobalt (<0.0030) %/Arsenic (<0.0050) %/Bismuth (<0.0050) %/Chromium (<0.0050) %/Molybdenum (<0.0050) %/Titanium (<0.0050) %/Silver (<0.0060) %/Lead (<0.0070) %/Selenium (<0.0100) %/Tin (<0.0100) %/Antimony (<0.0200) %/Phosphorus (<0.0200) %/Cadmium (0.0003) %/Nitrogen (0.0006) %/Oxygen (0.0008) %/Sulfur (0.0009) %/Niobium (0.0012) %/Carbon (0.0017) %/Boron (0.003) %/Copper (96.5) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction