missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon Steel, Rephosphorized and Resulfurized, AISI 12L14, G12144, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Carbon Steel, Rephosphorized and Resulfurized, AISI 12L14, G12144, 100g
Specifications
Specifications
| Chemical Name or Material | Silicon/Boron/Niobium/Titanium/Zirconium/Aluminium/Tin/Bismuth/Arsenic/Tungsten/Vanadium/Antimony/Molybdenum/Cobalt/Nitrogen/Oxygen/Copper/Nickel/Chromium/Carbon/Phosphorus/Lead/Sulfur/Manganese/Iron |
| CAS | 7439-89-6 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Silicon -48 ppm/Boron -11 ppm/Niobium -5 ppm/Titanium -4 ppm/Zirconium 0.0009 %/Aluminium 0.0013 %/Tin 0.0013 %/Bismuth 0.0024 %/Arsenic 0.0027 %/Tungsten 0.003 %/Vanadium 0.0032 %/Antimony 0.0033 %/Molybdenum 0.0034 %/Cobalt 0.0035 %/Nitrogen 0.006 %/Oxygen 0.012 %/Copper 0.0134 %/Nickel 0.0177 %/Chromium 0.056 %/Carbon 0.063 %/Phosphorus 0.071 %/Lead 0.29 %/Sulfur 0.32 %/Manganese 1.02 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction