missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alloy Steel, Nitriding Steel (135), Nitralloy 135M, K24065, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Alloy Steel, Nitriding Steel (135), Nitralloy 135M, K24065, 100g
Specifications
Specifications
| Chemical Name or Material | Boron/Calcium/Zirconium/Niobium/Antimony/Tungsten/Vanadium/Nitrogen/Titanium/Arsenic/Cobalt/Phosphorus/Tin/Nickel/Copper/Molybdenum/Silicon/Carbon/Manganese/Aluminium/Chromium/Lead/Oxygen/Magnesium/Zinc/Bismuth/Tantalum |
| CAS | 7439-89-6 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Boron 0.0006 %/Calcium 0.0007 %/Zirconium 0.0011 %/Niobium 0.002 %/Antimony 0.004 %/Tungsten 0.004 %/Vanadium 0.004 %/Nitrogen 0.0044 %/Titanium 0.0044 %/Arsenic 0.006 %/Cobalt 0.007 %/Phosphorus 0.011 %/Tin 0.011 %/Nickel 0.156 %/Copper 0.214 %/Molybdenum 0.32 %/Silicon 0.349 %/Carbon 0.425 %/Manganese 0.58 %/Aluminium 0.92 %/Chromium 1.63 %/Lead (0.0003) %/Oxygen (0.0006) %/Magnesium (0.002) %/Zinc (0.002) %/Bismuth <0.002 %/Tantalum <0.005 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction