missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alloy Steel, Cr/Mo, AISI 4140, G41400, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Alloy Steel, Cr/Mo, AISI 4140, G41400, 100g
Specifications
Specifications
| Chemical Name or Material | Oxygen/Niobium/Lead/Zirconium/Boron/Titanium/Tungsten/Vanadium/Antimony/Arsenic/Phosphorus/Cobalt/Tin/Nitrogen/Sulfur/Aluminium/Nickel/Molybdenum/Silicon/Copper/Carbon/Manganese/Chromium/Zinc/Magnesium/Iron |
| CAS | 7439-89-6 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Oxygen -40 ppm/Niobium -20 ppm/Lead -10 ppm/Zirconium -10 ppm/Boron -5 ppm/Titanium 0.0009 %/Tungsten 0.003 %/Vanadium 0.003 %/Antimony 0.004 %/Arsenic 0.005 %/Phosphorus 0.008 %/Cobalt 0.009 %/Tin 0.01 %/Nitrogen 0.011 %/Sulfur 0.022 %/Aluminium 0.031 %/Nickel 0.14 %/Molybdenum 0.17 %/Silicon 0.22 %/Copper 0.23 %/Carbon 0.401 %/Manganese 0.9 %/Chromium 1.1 %/Zinc (<30) ppm/Magnesium (<50) ppm/Iron (97.3) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction