missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alloy Steel, 11⁄4Cr 1⁄2Mo, A182 Grade F11, K11572, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Alloy Steel, 11⁄4Cr 1⁄2Mo, A182 Grade F11, K11572, 100g
Specifications
Specifications
| Chemical Name or Material | Boron/Oxygen/Niobium/Titanium/Vanadium/Arsenic/Nitrogen/Cobalt/Phosphorus/Tin/Aluminium/Nickel/Carbon/Copper/Manganese/Molybdenum/Silicon/Chromium/Calcium/Lead/Zirconium/Antimony/Tungsten |
| CAS | 7439-89-6 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Boron 0.0004 %/Oxygen 0.0016 %/Niobium 0.002 %/Titanium 0.003 %/Vanadium 0.005 %/Arsenic 0.008 %/Nitrogen 0.0093 %/Cobalt 0.011 %/Phosphorus 0.012 %/Tin 0.017 %/Aluminium 0.029 %/Nickel 0.102 %/Carbon 0.115 %/Copper 0.271 %/Manganese 0.44 %/Molybdenum 0.52 %/Silicon 0.56 %/Chromium 1.09 %/Calcium (0.0004) %/Lead (0.001) %/Zirconium (0.002) %/Antimony (0.005) %/Tungsten (0.005) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction