missing translation for 'onlineSavingsMsg'
Learn More

10058-F4, MedChemExpress

Product Code. 16465298
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16465298 10 mg 10mg
16421507 50 mg 50mg
16418017 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16465298 Supplier Medchem Express Supplier No. HY12702
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 10058-F4 is a c-Myc inhibitor that prevents c-Myc-Max dimerization and transactivation of c-Myc target gene expression.

10058-F4 is a c-Myc inhibitor that prevents c-Myc-Max dimerization and transactivation of c-Myc target gene expression.

Specifications

Chemical Name or Material 10058-F4
Physical Form Solid
CAS 403811-55-2
Color Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C12H11NOS2
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : ≥ 41 mg/mL (164.43 mM)
SMILES O=C1NC(S/C1=C/C2=CC=C(CC)C=C2)=S
Molecular Weight (g/mol) 249.35
Formula Weight 249.35
Percent Purity 99.18%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.