missing translation for 'onlineSavingsMsg'
Learn More

10058-F4, MedChemExpress

Product Code. 16418017
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16418017

Brand: Medchem Express HY12702

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 10058-F4 is a c-Myc inhibitor that prevents c-Myc-Max dimerization and transactivation of c-Myc target gene expression.

10058-F4 is a c-Myc inhibitor that prevents c-Myc-Max dimerization and transactivation of c-Myc target gene expression.

Specifications

Chemical Name or Material 10058-F4
Physical Form Solid
CAS 403811-55-2
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C12H11NOS2
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : ≥ 41 mg/mL (164.43 mM)
SMILES O=C1NC(S/C1=C/C2=CC=C(CC)C=C2)=S
Molecular Weight (g/mol) 249.35
Formula Weight 249.35
Percent Purity 99.18%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.