missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1000 NTU Ratio Turbidity Standard, Reagecon
Suspension of extremely accurate (± 1%) polymer microspheres with guaranteed shelf life stability even after opening the bottle.
£180.00 - £410.00
Specifications
For Use With (Equipment) | Ratio Instruments |
---|---|
Accuracy | Accuracy ± 1% NTU |
Turbidity | 1000 NTU ± 1% |
Product Code | Brand | Description | Volume (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Volume (Metric) | Price | Quantity & Availability | ||||
11536593
|
Reagecon
CRSR-1000-100 |
Liquid format in an HDPE Bottle | 100 mL |
£180.00
100mL |
Estimated Shipment: 20-05-2024
Log in to see stock. |
||||
12736935
|
Reagecon
CRSR-1000-500 |
Liquid format in an HDPE Bottle GL45 | 500 mL |
£410.00
500mL |
Estimated Shipment: 17-05-2024
Log in to see stock. |
||||
Description
- Turbidity Reference Standard, Turbidity Reference Material.
- Designed and formulated to overcome the adverse safety, accuracy and stability issues experienced with other turbidity standards, primarily formazin-based standards.
- Certified
- NIST traceable
- Non-toxic, non-carcinogenic
- Highly accurate
- Stable
- EPA approved
Specifications
Ratio Instruments | |
Accuracy ± 1% NTU | |
1000 NTU ± 1% |