Learn More
Eppendorf™ 1000μL Polypropylene 96-Well Deep Well PCR Plates
GREENER_CHOICE
Accelerate your manual and automated work processes with these 96-well polypropylene plates. Eppendorf™ 1000μL Polypropylene 96-Well Deep Well PCR Plates allow high speed pipetting without splashing, excellent mixing and high speed centrifugation.
Brand:  Eppendorf™ 0030509200
Product Code. 10360665
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • These polypropylene plates combine a new well design with a sophisticated color coding system for ease of reading and orientation
  • Fulfill all current SBS, IVD, EN ISO 9001/2/3 and EN 13485 guidelines
  • Recovermax - concaved round bottom well shape with smoothed edges
  • Optitrack - color framed plates with alphanumeric labeling
  • High chemical resistance
  • G-safe - highest centrifugation stability
  • LoBind - free of surface coating e.g., silicone
  • Free from DNA, DNase, RNase and PCR* inhibitor (PCR* clean)
  • LoBind Protein - minimum protein loss (less than 3% BSA 1μg/mL)
  • LoBind DNA - less than 1% loss of DNA/RNA at low concentrations and critical conditions (high salt, short DNA fragments)
  • Autoclavable
  • Optionally available with barcode