missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Tau K18/Tau PHF Core Protein

Product Code. 18741838
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 18741838

Brand: R&D Systems™ SP496100

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human Tau K18/Tau PHF Core Protein is derived from E. coli. The Recombinant Human Tau K18/Tau PHF Core Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P10636-8
For Use With (Application) Bioactivity
Gene ID (Entrez) 4137
Molecular Weight (g/mol) 14 kDa
Quantity 100 μg
Source E. coli-derived human Tau protein Leu243-Glu372
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 24 months from date of receipt, -70 °C as supplied. 6 months, -70 °C under sterile conditions after opening.
Regulatory Status RUO
Specific Reactivity Tau
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Buffer X mg/mL (X μm) in PBS pH 7.4
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie« Blue stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.