missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat GABAB R2 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB104561-100
This item is not returnable.
View return policy
Description
GABA-B R2 Monoclonal specifically detects GABA-B R2 in Mouse, Rat samples. It is validated for Western Blot, Simple Western.
Specifications
| GABA-B R2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| G protein-coupled receptor 51, GABABR2, GABA-BR2, GABA-B-R2, GABABR2GABA-B receptor 2, GABBR2, gamma-aminobutyric acid (GABA) B receptor, 2, gamma-aminobutyric acid type B receptor subunit 2, gb2, GPR51, GPR51GABA-B receptor, R2 subunit, GPRC3B, GPRC3BFLJ36928, G-protein coupled receptor 51, HG20HRIHFB2099 | |
| Mouse myeloma cell line NS0-derived recombinant rat GABAB R2, Trp41-Ser482, Accession # O88871 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG |
| Western Blot | |
| 2605F | |
| Western Blot 1 ug/mL, Simple Western 20 ug/mL | |
| O88871 | |
| Rabbit | |
| Protein A or G purified from egg yolks | |
| RUO | |
| 9568 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction