missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CXCL1/GRO alpha/KC/CINC-1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB4532-100
This item is not returnable.
View return policy
Description
CXCL1/GRO alpha/KC/CINC-1 Monoclonal specifically detects CXCL1/GRO alpha/KC/CINC-1 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| CXCL1/GRO alpha/KC/CINC-1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| CINC1, CINC-1, FSP, GRO alpha, GRO1, GROa, KC, MGSA, MGSA-a, MGSA-alpha, NAP-3, SCYB1 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects mouse CXCL1/GRO alpha/KC/CINC-1 in direct ELISAs. | |
| Mouse | |
| Lyophilized |
| Flow Cytometry | |
| 1174A | |
| Intracellular Staining by Flow Cytometry 0.25-1 μg/106 cells | |
| P12850 | |
| Cxcl1 | |
| E. coli-derived recombinant mouse CXCL1/GRO alpha/KC/CINC-1, Asn29-Lys96, Accession No. P12850 | |
| 100 μg | |
| 2919.0 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction