missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TMED1 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB2243S-100UG
This item is not returnable.
View return policy
Description
TMED1 Monoclonal specifically detects TMED1 in Human samples. It is validated for Flow Cytometry.
Specifications
| TMED1 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| IL1RL1-Binding Protein, Il1rl1l, IL1RL1LG, IL-1RL1LG, IL1RL1LGIL1RL1-binding protein, Interleukin 1 Receptor-Like 1 Ligand, Interleukin-1 receptor-like 1 ligand, Ly84l, MGC1270, P24g1, P24gamma1, Putative T1/ST2 receptor-binding protein, ST2L, T1/ST2 receptor binding protein, Tp24, transmembrane emp24 domain containing 1, transmembrane emp24 domain-containing protein 1, transmembrane emp24 protein transport domain containing 1 | |
| Human embryonic kidney cell, HEK293 derived human TMED1, Met1-Asn194, Accession # Q13445 | |
| 100 μg | |
| 11018 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 1009527 | |
| Flow Cytometry 0.25-1 μg/106 cells | |
| Q13445 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human TMED1 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction